Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome b245 Light Chain Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3879R

Description
Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.Specifications
Cytochrome b245 Light Chain | |
Polyclonal | |
Unconjugated | |
CYBA | |
b558; CYBA; cytochrome b light chain; Cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; cytochrome b-245 alpha chain; Cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; Cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; Neutrophil cytochrome b 22 kDa polypeptide; nmf333; p22 phagocyte B-cytochrome; p22 phox; p22phox; p22-phox; Superoxide-generating NADPH oxidase light chain subunit | |
Rabbit | |
Protein A | |
RUO | |
1535 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P13498 | |
CYBA | |
KLH conjugated synthetic peptide derived from human CK18 Cytochrome b245 Light Chain/p22phox. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction