Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome b5 Outer Mitochondrial Membrane Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297403AF350
Description
Cytochrome b5 Outer Mitochondrial Membrane Polyclonal specifically detects Cytochrome b5 Outer Mitochondrial Membrane in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytochrome b5 Outer Mitochondrial Membrane | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CYB5M, CYB5-M, CYPB5M, Cytochrome b5 outer mitochondrial membrane isoform, cytochrome b5 type B, cytochrome b5 type B (outer mitochondrial membrane), DKFZp686M0619, OMB5, outer mitochondrial membrane cytochrome b5, type 2 cyt-b5 | |
Produced in rabbits immunized with E. coli-derived Human Cytochrome b5 Outer Mitochondrial Membrane fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
80777 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction