Learn More
Invitrogen™ Cytochrome B5 Polyclonal Antibody

Description
Recommended positive controls: Cytochrome b5 tansfected 293T cell. Predicted reactivity: Mouse (90%), Rat (90%), Xenopus laevis (81%), Dog (96%), Pig (93%), Chicken (82%), Sheep (93%), Rhesus Monkey (97%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Cytochrome B5 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CYB5A |
| Gene Accession No. | P00167 |
| Gene Alias | 0610009N12Rik; Cyb5; CYB5A; cytochrome b5; cytochrome b-5; cytochrome b5 type A; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5 |
| Gene Symbols | CYB5A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.