Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome b5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986339
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6339 Supplier Bioss Supplier No. BS5115R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber′s hereditary optic neuropathy and with myopathy.

Specifications

Antigen Cytochrome b5
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CYB5A
Gene Accession No. P00167
Gene Alias 0610009N12Rik; CYB5; Cyb5a; Cytochrome b5; cytochrome b-5; cytochrome b5 type A; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5
Gene Symbols CYB5A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Cytochrome b5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1528
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.