Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome c Antibody (7H8.2C12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 32 publications
Supplier: Novus Biologicals NB10055775
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Drosophila, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
Cytochrome c | |
Monoclonal | |
0.5mg/mL | |
Western Blot 0.05-0.5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular) | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
An approx. 15 kDa band is observed. This antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached). | |
Store at 4C. Do not freeze. | |
IgG2b κ |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Flow Cytometry | |
7H8.2C12 | |
Biotin | |
P99999 | |
CYCS | |
Synthetic peptides corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYT were used as the immunogen (Jemmerson et al. 1991). The antibody recognizes an epitope within amino acids 93-104 of pigeon cytochrome C based on competitive ELISA results (Jemmerson et al. 1991). | |
0.1 mL | |
Apoptosis, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
54205 | |
Human, Mouse, Rat, Drosophila, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction