Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome c Antibody (CTC05), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311456PCP
Description
Cytochrome c Monoclonal antibody specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Flow CytometrySpecifications
Cytochrome c | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon | |
Purified |
Flow Cytometry | |
CTC05 | |
Flow Cytometry | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Recombinant cytochrome c protein (Uniprot: P99999) | |
0.1 mL | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
54205 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction