Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome c Antibody (SPM389) - IHC-Prediluted, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP248147
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Avian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytochrome c | |
Monoclonal | |
Unconjugated | |
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CYCS | |
Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon cytochrome c (Uniprot: P99999) | |
Protein A or G purified | |
RUO | |
Primary | |
It recognizes an epitope within amino acids 93-104 of pigeon Cytochrome C, a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of cytochrome c, thereby blocking the apoptotic process. Overexpression of Bax has been shown to induce the release of cytochrome c and to induce cell death. The release of cytochrome c from the mitochondria is thought to trigger an apoptotic cascade, whereby Apaf-1 binds to Apaf-3 (caspase-9) in a cytochrome c-dependent manner, leading to caspase-9 cleavage of caspase-3. This monoclonal antibody recognizes total cytochrome C which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached). | |
Store at 4C. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
SPM389 | |
Immunohistochemistry-Paraffin | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Mouse | |
15 kDa | |
7 mL | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
54205 | |
Human, Mouse, Rat, Amphibian, Avian, Canine, Drosophila, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction