Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome C Monoclonal Antibody (CTC03 (2B5)), Invitrogen™

Catalog No. PIMA511823
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511823 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511823 Supplier Thermo Scientific Supplier No. MA511823

Mouse Monoclonal Antibody

MA5-11823 targets Cytochrome c in ELISA, FACS, immunofluorescence and immunoprecipitation applications and shows reactivity with Bovine, Canine, Equine, Mouse and Human samples. MA5-11823 is not suitable for Western blotting applications. This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.

Cytochrome C Monoclonal antibody specifically detects Cytochrome C in Bovine, Canine, Equine, Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone CTC03 (2B5)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Cycs
Gene Accession No. P00004, P00011, P62894, P62897, P99999
Gene Alias CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4
Gene Symbols Cycs
Host Species Mouse
Immunogen Equine cytochrome c
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100053958, 13063, 475258, 510767, 54205
Target Species Bovine, Canine, Equine, Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.