Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome c Mouse anti-Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon, Clone: rCYCS/1010, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cytochrome c |
---|---|
Clone | rCYCS/1010 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
Cytochrome c | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
54205 | |
IgG1 κ | |
Protein A or G purified |
rCYCS/1010 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Amphibian, Canine, Drosophila, Equine, Pigeon | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Synthetic peptides corresponding to amino acid 1-80, 81-104 and 66-104 of pigeon Cytochrome c | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title