Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome C Oxidase subunit 6c Antibody (OTI4B11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245645
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
Cytochrome C Oxidase subunit 6c Monoclonal specifically detects Cytochrome C Oxidase subunit 6c in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
Cytochrome C Oxidase subunit 6c | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
Cytochrome c oxidase polypeptide VIc, cytochrome c oxidase subunit 6C, cytochrome c oxidase subunit VIc, cytochrome c oxidase subunit VIc preprotein | |
Mouse | |
8.6 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI4B11 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
COX6C | |
Full length human recombinant protein of human COX6C(NP_004365) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
1345 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction