Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytochrome C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528856 Supplier Invitrogen™ Supplier No. PA528856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, K562, THP-1, HL-60, Mouse brain, PC-12, CYCS shRNA-transfected 293T, 293T, SCC-25. Predicted reactivity: Mouse (91%), Rat (91%), Xenopus laevis (85%), Dog (89%), Pig (90%), Chicken (87%), Chimpanzee (100%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Cycs
Gene Accession No. P62897, P62898, P99999
Gene Alias CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4
Gene Symbols Cycs
Host Species Rabbit
Immunogen Full length human Cytochrome C Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13063, 25309, 54205
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.