Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytochrome C1 Monoclonal Antibody (27E12D11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA550277
Description
Cytochrome C1 Monoclonal Antibody for Western Blot, Flow, ELISA
This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.
Specifications
Cytochrome C1 | |
Monoclonal | |
1.421 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P08574, Q9D0M3 | |
CYC1 | |
Recombinant Human Cytochrome c1 protein (85-325AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
27E12D11 | |
Unconjugated | |
CYC1 | |
2610002H19Rik; AA408921; Complex III subunit 4; complex III subunit IV; Cyc1; Cytochrome b-c1 complex subunit 4; cytochrome c1; cytochrome c-1; cytochrome c1, heme protein, mitochondrial; MC3DN6; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4 | |
Mouse | |
Protein G | |
RUO | |
1537, 66445 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction