Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome C1 Polyclonal Antibody
Cytochrome C1 Polyclonal Antibody
Supplier: Thermo Scientific PA527490
Description
Cytochrome C1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
Cytochrome C1 | |
Polyclonal | |
Unconjugated | |
CYC1 | |
2610002H19Rik; AA408921; Complex III subunit 4; complex III subunit IV; Cyc1; Cytochrome b-c1 complex subunit 4; cytochrome c1; cytochrome c-1; cytochrome c1, heme protein, mitochondrial; MC3DN6; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4 | |
Rabbit | |
Protein A | |
RUO | |
1537, 66445 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
P08574, Q9D0M3 | |
CYC1 | |
Recombinant fragment corresponding to a region within amino acids 54 and 301 of Human Cytochrome C1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction