Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytochrome C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551550 Supplier Invitrogen™ Supplier No. PA551550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSEAARAANN GALPPDLSYI VRARHGGEDY VFSLLTGYCE PPTGVSLREG LYFNPYFPGQ AIAMAPPIYT DVLEFDDGTP ATMSQIAKDV CTFLRWASEP EHDHRKRMGL KMLMMMALLV PLVYTIKRHK WSVLKSRKLA YR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome C1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYC1
Gene Accession No. P08574, Q9D0M3
Gene Alias 2610002H19Rik; AA408921; Complex III subunit 4; complex III subunit IV; Cyc1; Cytochrome b-c1 complex subunit 4; cytochrome c1; cytochrome c-1; cytochrome c1, heme protein, mitochondrial; MC3DN6; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4
Gene Symbols CYC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cytochrome C1. Recombinant protein control fragment (Product #RP-99401).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1537, 300047, 66445
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.