Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytochrome C1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101998
Description
Antibody detects endogenous levels of total CYC1.
This is the heme-containing component of the cytochrome b-c1 complex, which accepts electrons from Rieske protein and transfers electrons to cytochrome c in the mitochondrial respiratory chain.
Specifications
Cytochrome C1 | |
Polyclonal | |
Unconjugated | |
CYC1 | |
2610002H19Rik; AA408921; Complex III subunit 4; complex III subunit IV; Cyc1; Cytochrome b-c1 complex subunit 4; cytochrome c1; cytochrome c-1; cytochrome c1, heme protein, mitochondrial; MC3DN6; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1537, 300047, 66445 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P08574, Q9D0M3 | |
CYC1 | |
A synthesized peptide derived from human CYC1(Accession P08574), corresponding to amino acid residues A131-N181. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction