Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 17A1 Antibody (OTI3F12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270542B
Description
Cytochrome P450 17A1 Monoclonal specifically detects Cytochrome P450 17A1 in Human samples. It is validated for Western Blot.Specifications
Cytochrome P450 17A1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CYP17A1 | |
Full length recombinant protein of human Cytochrome P450 17A1 (NP_000093) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI3F12 | |
Western Blot | |
CPT7, cyp17, CYP17A1, cytochrome p450 XVIIA1, cytochrome P450, family 17, subfamily A, polypeptide 1, cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenalhyperplasia, Cytochrome P450c17, Cytochrome P450-C17, EC 1.14.99, P450C17, P450C17CYPXVII, S17AH | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
1586.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction