Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 1A1/1A2 Mouse anti-Human, Mouse, Rat, Primate, Clone: MC1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cytochrome P450 1A1/1A2 |
---|---|
Clone | MC1 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Cytochrome P450 1A1/1A2 Monoclonal specifically detects Cytochrome P450 1A1/1A2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cytochrome P450 1A1/1A2 | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Breast Cancer, Lipid and Metabolism | |
10 mM PBS with 0.05% BSA | |
1543 | |
IgG1 κ | |
Protein A or G purified |
MC1 | |
Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
AHH, AHRR, aryl hydrocarbon hydroxylase, CP11, CYP1, CYPIA1, cytochrome P1-450, dioxin-inducible, cytochrome P450 1A1, Cytochrome P450 form 6, cytochrome P450, family 1, subfamily A, polypeptide 1, Cytochrome P450-C, Cytochrome P450-P1, EC 1.14.14.1, flavoprotein-linked monooxygenase, P1-450, P450-C, P450DX, subfamily I (aromatic compound-inducible), polypeptide 1, xenobiotic monooxygenase | |
3-methylcholanthrene induced rat cytochrome P450 protein. | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title