Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 1A2 Antibody (D15), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001490C
Description
Cytochrome P450 1A2 Monoclonal specifically detects Cytochrome P450 1A2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Cytochrome P450 1A2 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP1A2 | |
MC1a (Preparation C31B4, rat liver cytochrome P4501A2) | |
0.1 mL | |
Cancer, Lipid and Metabolism | |
1544 | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
D15 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
aryl hydrocarbon hydroxylase, CP12, CYPIA2, Cytochrome P(3)450, cytochrome P450 1A2, Cytochrome P450 4, cytochrome P450, family 1, subfamily A, polypeptide 2, cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2, Cytochrome P450-P3, dioxin-inducible P3-450, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P3-450, P450 form 4, P450(PA), xenobiotic monooxygenase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
D15 cross reacts with CYP1A1 & CYP1A2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction