Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cytochrome P450 26A1, Mouse, Unlabeled, Clone: F27P6A1,

Catalog No. MABS1319MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1319MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1319MI Supplier MilliporeSigma™ Supplier No. MABS1319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 26A1 (UniProt O43174; also known as Cytochrome P450 retinoic acid-inactivating 1, Cytochrome P450RAI, hP450RAI, Retinoic acid 4-hydroxylase, Retinoic acid-metabolizing cytochrome) is encoded by the CYP26A1 (also known as CYP26, P450RAI1) gene (Gene ID 1592) in human. CYP26A1 is a retinoic acid (RA) metabolizing enzyme that is mainly located in the endoplasmic reticulum membranes and is involved in vitamin A metabolism. It can generate several hydroxylated forms of RA, including 4-OH-RA, 4-oxo-RA and 18-OH-RA. RA is a tight binding ligand for CYP26A1 with lownm binding affinity. Higher expression of CYP26A1 is observed in the adult liver, heart, pituitary gland, placenta, and various regions of the brain. CYP26A1 activity is important for the maintenance of pregnancy, especially during the process of blastocyst implantation. Elevated CYP26A1 expression and RA catabolic activity have been detected in breast epithelial adenocarcinoma cells in culture, in leukemic cells from patients with acute promyelocytic leukemia, and in cells derived from squamous cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 26A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone F27P6A1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CYP26A1, CYP26, P450RAI1
Gene Accession No. O43174
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to a sequence near the C-terminus of human cytochrome P450 26A1 (Kumarakulasingham, M., et al. (2005). Clin. Cancer Res. 11(10):3758-3765).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000774
Test Specificity Target band specificity was confirmed by Western blotting of lysate from cytochrome P450 26A1-overexpressing human embryonic kidney cells (Brown, G.T., et al. (2014). PLoS One. 9(3):e90776).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.