Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 26B1 Antibody (T5P3G2*A3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250195AF594
Description
Cytochrome P450 26B1 Monoclonal specifically detects Cytochrome P450 26B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytochrome P450 26B1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CYP26B1 | |
Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
T5P3G2*A3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
Mouse | |
Protein A purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
56603.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction