Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 26B1 Mouse anti-Human, Mouse, Rat, Clone: T5P3G2/A3, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50
Specifications
Antigen | Cytochrome P450 26B1 |
---|---|
Clone | T5P3G2/A3 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBPY1950025
![]() |
Novus Biologicals
NBP2501950.025ML |
0.025 mL |
Each of 1 for $243.50
|
|
|||||
Description
Cytochrome P450 26B1 Monoclonal specifically detects Cytochrome P450 26B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytochrome P450 26B1 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
CYP26B1 | |
IgG1 κ | |
Protein G purified |
T5P3G2/A3 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
56603 | |
Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title