Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cytochrome P450 26B1, Mouse, Unlabeled, Clone: 4B7.2,

Catalog No. MABS497MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS497MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS497MI Supplier MilliporeSigma™ Supplier No. MABS497
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytochrome P450 26B1 (UniProt Q9NR63; also known as Cytochrome P450 26A2, Cytochrome P450 retinoic acid-inactivating 2, Cytochrome P450, subfamily XXVIB, polypeptide 1, Cytochrome P450RAI-2, Retinoic acid-metabolizing cytochrome) is encoded by the CYP26B1 (also known as CYP26A2, P450RAI2, RHFCA) gene (Gene ID 56603) in human. Cytochromes P450 (CYP) proteins are primarily membrane-associated oxidases located either in the inner membrane of mitochondria or in the endoplasmic reticulum. In addition to processing endogenous substrates, CYPs also metabolize exogenous drugs and potentially toxic chemicals. The human CYP superfamily consists of 57 genes and more than 59 pseudogenes divided into 18 families and 43 subfamilies. The CYP26 subfamily of enzymes (CYP26A1, CYP26B1/CYP26A2, CYP26C1) are retinoic acid hydroxylases responsible for the inactivation of all-trans-retinoic acid to hydroxylated forms, such as 4-oxo-, 4-OH-, and 18-OH-all-trans-retinoic acid. CYP26B1 is overexpressed in colorectal cancer (CRC) cells, high CYP26B1 expression level in CRC patients is associated with poor prognosis. CYP26B1 mutations are found in radiohumeral fusions with other skeletal and craniofacial anomalies (RHFCA), a disease characterized by craniofacial malformations, occipital encephalocele, radiohumeral fusions, oligodactyly, advanced osseous maturation, and calvarial mineralization defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 26B1
Applications Western Blot
Classification Monoclonal
Clone 4B7.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene CYP26B1, CYP26A2, P450RAI2, RHFCA
Gene Accession No. Q9NR63
Host Species Mouse
Immunogen GST-tagged recombinant internal fragment from the C-terminal half of human Cytochrome P450 26B1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_063938
Test Specificity Expected to react with all three spliced isoforms of human CYP26B1 reported by UniProt (Q9NR63).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.