Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2B1, 2B2 Antibody (b/e3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250201AF405
Description
Cytochrome P450 2B1, 2B2 Monoclonal specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Cytochrome P450 2B1, 2B2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rat liver cytochromes p450 2B1 and 2B2 | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
Clone b/e3 inhibits PROD activity in liver microsomes of PB-treated rats and inhibits ECOD activity in liver microsomes of PB-treated but not 3MC-treated rats. The CYP2B1 antibody recognizes the major barbiturate-inducible P450s in mouse and rat. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
b/e3 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction