Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2B1, 2B2 Antibody (b/e4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250202B
Description
Cytochrome P450 2B1, 2B2 Monoclonal specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, ELISA.Specifications
Cytochrome P450 2B1, 2B2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Rat liver cytochromes P4502B1 and 2B2 | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
Mouse, Rat, Canine, Primate | |
Purified |
Western Blot, ELISA | |
b/e4 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction