Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2B6 Antibody (OTI3D5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270526MFV450
Description
Cytochrome P450 2B6 Monoclonal antibody specifically detects Cytochrome P450 2B6 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
Cytochrome P450 2B6 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Flow Cytometry | |
OTI3D5 | |
Flow Cytometry | |
CPB6, CYP2B, CYP2B7, CYP2B7P, CYPIIB6cytochrome P450, subfamily IIB (phenobarbital-inducible), cytochrome P450 2B6, Cytochrome P450 IIB1, cytochrome P450, family 2, subfamily B, cytochrome P450, family 2, subfamily B, polypeptide 6, cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6, EC 1.14.14.1, IIB1, P450 | |
Full length human recombinant protein of human CYP2B6(NP_000758) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
1555 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction