Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2C8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Cytochrome P450 2C8 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cytochrome P450 2C8 Polyclonal specifically detects Cytochrome P450 2C8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytochrome P450 2C8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
CPC8, CYPIIC8, cytochrome P450 2C8, Cytochrome P450 form 1, Cytochrome P450 IIC2, Cytochrome P450 MP-12, Cytochrome P450 MP-20, cytochrome P450, family 2, subfamily C, polypeptide 8, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, MP-12/MP-20, P450 form 1, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
CYP2C8 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
1558 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KRICAGEGLARMELFLFLTTILQNFNLKSVDDLKNLNTTAVTKGIVSLPPSYQICFIPV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title