Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2C9 Antibody (OTI1D7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270528R
Description
Cytochrome P450 2C9 Monoclonal specifically detects Cytochrome P450 2C9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Cytochrome P450 2C9 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP2C9 | |
Full length recombinant protein of human Cytochrome P450 2C9 (NP_000762) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
(R)-limonene 6-monooxygenase, (S)-limonene 6-monooxygenase, (S)-limonene 7-monooxygenase, CPC9, CYP2C, CYP2C10, CYPIIC9, cytochrome P450 2C9, Cytochrome P450 MP-4, Cytochrome P450 MP-8, Cytochrome P450 PB-1, cytochrome P-450 S-mephenytoin 4-hydroxylase, cytochrome P450, family 2, subfamily C, polypeptide 9, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 9, Cytochrome P-450MP, EC 1.14.13.48, EC 1.14.13.49, EC 1.14.13.80, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC149605, MGC88320, microsomal monooxygenase, P450IIC9, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
1559.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction