Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2C9 Antibody (SR1875), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321949100UL
Description
Cytochrome P450 2C9 Monoclonal antibody specifically detects Cytochrome P450 2C9 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
Cytochrome P450 2C9 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1875 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
(R)-limonene 6-monooxygenase, (S)-limonene 6-monooxygenase, (S)-limonene 7-monooxygenase, CPC9, CYP2C, CYP2C10, CYPIIC9, cytochrome P450 2C9, Cytochrome P450 MP-4, Cytochrome P450 MP-8, Cytochrome P450 PB-1, cytochrome P-450 S-mephenytoin 4-hydroxylase, cytochrome P450, family 2, subfamily C, polypeptide 9, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 9, Cytochrome P-450MP, EC 1.14.13.48, EC 1.14.13.49, EC 1.14.13.80, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC149605, MGC88320, microsomal monooxygenase, P450IIC9, S-mephenytoin 4-hydroxylase, xenobiotic monooxygenase | |
A synthesized peptide derived from human Cytochrome P450 2C9 (Uniprot #: P11712) | |
100 μg | |
Lipid and Metabolism | |
1559 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction