Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 2E1 Antibody (M12P4H2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308831AF647
Description
Cytochrome P450 2E1 Monoclonal antibody specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Cytochrome P450 2E1 | |
Monoclonal | |
Alexa Fluor 647 | |
4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase | |
Ovalbumin-conjugated synthetic peptide HIGFGCIPPR (C-terminal sequence) (Uniprot: P05181) | |
100 μg | |
Lipid and Metabolism | |
1571 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
M12P4H2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction