Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome p450 2J2 Antibody (OTI5C9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270533F
Description
Cytochrome p450 2J2 Monoclonal specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Immunohistochemistry.Specifications
Cytochrome p450 2J2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
CYP2J2 | |
Full length recombinant protein of human Cytochrome p450 2J2 (NP_000766) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI5C9 | |
Immunohistochemistry | |
Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
1573.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction