Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Cytochrome P450 3A5 Antibody (F18P3B6), mFluor Violet 610 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250209MFV610
Description
Cytochrome P450 3A5 Monoclonal antibody specifically detects Cytochrome P450 3A5 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Cytochrome P450 3A5 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, ELISA, Immunohistochemistry | |
| F18P3B6 | |
| Western Blot, ELISA, Immunohistochemistry | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Ovalbumin-conjugated synthetic peptide DSRDGTLSGE (C-terminal sequence) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1577 | |
| Store at 4C in the dark. | |
| IgG1 κ | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction