Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 4A Antibody (M25-P2A10), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250212AF350
Description
Cytochrome P450 4A Monoclonal specifically detects Cytochrome P450 4A in Human, Mouse, Rat, Drosophila, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Cytochrome P450 4A | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CYP4A11 | |
Peptide Sequence - KNGIHLRLR (amino acids 499 - 507) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Drosophila, Xenopus species | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
M25-P2A10 | |
Western Blot, ELISA, Immunohistochemistry | |
20-HETE synthase, CP4Y, CYP4A2, cytochrome P450 4A11, cytochrome P450, family 4, subfamily A, polypeptide 11, cytochrome P450, subfamily IVA, polypeptide 11, cytochrome P-450HK-omega, cytochrome P450HL-omega, fatty acid omega-hydroxylase, lauric acid omega-hydroxylase, P450HL-omega | |
Mouse | |
Protein A purified | |
Lipid and Metabolism | |
1579.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction