Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytochrome P450 4A1, 2C11 Antibody (Clo1), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250211UV
Description
Cytochrome P450 4A1, 2C11 Monoclonal specifically detects Cytochrome P450 4A1, 2C11 in Rat samples. It is validated for Western Blot, ELISA.Specifications
Cytochrome P450 4A1, 2C11 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, ELISA | |
Clo1 | |
Western Blot, ELISA | |
CYP4A1 | |
Rat liver cytochrome P450 2C11 and 4A1 | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction