Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytochrome P450 Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536232 Supplier Invitrogen™ Supplier No. PA536232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.

CYP2C9 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. CYP2C9 localizes to the endoplasmic reticulum and its expression is induced by rifampin. The enzyme is known to metabolize many xenobiotics, including phenytoin, tolbutamide, ibuprofen and S-warfarin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytochrome P450 Pan
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP2C9
Gene Accession No. P05181, P05182, P10632, P11712, P33260, P33261, Q05421
Gene Alias (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; (S)-mephenytoin hydroxylase associated cytochrome P450; cholesterol 25-hydroxylase; CPC8; CPC9; CPCI; CPCJ; CYP2C; CYP2C10; CYP2C17; CYP2C18; CYP2C19; CYP2C8; CYP2C9; CYPIIC17; CYPIIC18; CYPIIC19; CYPIIC8; CYPIIC9; Cytochrome P450 2C18; cytochrome P450 2C19; Cytochrome P450 2C8; cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 18; cytochrome P450 family 2 subfamily C member 19; cytochrome P450 family 2 subfamily C member 8; cytochrome P450 family 2 subfamily C member 9; cytochrome P450 form 1; cytochrome P-450 II C; Cytochrome P450 IIC2; cytochrome P450 MP-12; Cytochrome P450 MP-20; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; cytochrome P450-11A; Cytochrome P450-254C; cytochrome P450-6B/29C; cytochrome P-450MP; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; mephenytoin 4-hydroxylase; microsomal monooxygenase; MP-12/MP-20; P450 form 1; P450-6B/29C; P450C2C; P450IIC17; P450IIC19; P450IIC9; s-mephenytoin 4-hydroxylase; unspecific monooxygenase; xenobiotic monooxygenase
Gene Symbols CYP2C18, CYP2C19, CYP2C8, CYP2C9, CYP2E1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 100-150 of human CYP2C8.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13106, 1557, 1558, 1559, 1562, 1571, 25086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.