Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome P450 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992147
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2147 100 μg
50-199-2148 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2147 Supplier Rockland Immunochemicals Supplier No. 600401HB7
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human cytochrome p450. Blast analysis the immunogen sequence shows 83.3% identity with mouse, rat, hamster, and monkey.

This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.

Specifications

Antigen Cytochrome P450
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene CYP1A1
Gene Accession No. P00185, P04798
Gene Alias AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; CP11; CYP; CYP1; CYP1A1; Cyp1a-1; Cyp1a2; Cyp45c; Cypc45c; CYPIA1; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P450 1A1; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily a polypeptide 2; Cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, family 1, subfamily A, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; Cytochrome P450-C; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; Cytochrome P450-P1; dioxin-inducible; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P450; P450 form 6; P450-1; P450-C; P450DX; P450form6; P-450MC; P450MT2; P450-P1; xenobiotic monooxygenase
Gene Symbols CYP1A1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1543, 24296
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.