Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytoglobin Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300284MFV450
Description
Cytoglobin Polyclonal antibody specifically detects Cytoglobin in Human samples. It is validated for ELISASpecifications
Cytoglobin | |
Polyclonal | |
ELISA | |
cytoglobin, HGBHistoglobin, histoglobin, STAPHGb, Stellate cell activation-associated protein | |
Produced in rabbits immunized with purified, recombinant Human Cytoglobin (Accession#: AAH29798.1; Met1-Pro190) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
114757 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction