Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytoglobin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530957
Description
Recommended positive controls: IMR32, mouse heart, PC-12. Predicted reactivity: Human (99%), Mouse (94%), Rat (93%), Dog (94%), Sheep (96%), Rhesus Monkey (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002).
Specifications
| Cytoglobin | |
| Polyclonal | |
| Unconjugated | |
| CYGB | |
| 3110001K20Rik; Cygb; Cytoglobin; HGb; Histoglobin; Staap; STAP; stellate cell activation associated protein; stellate cell activation-associated protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114757, 114886, 170520 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WWM9, Q921A4, Q9CX80 | |
| CYGB | |
| Recombinant protein encompassing a sequence within the center region of human Cytoglobin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction