Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytoglobin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553637 Supplier Invitrogen™ Supplier No. PA553637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AILVRFFVNF PSAKQYFSQF KHMEDPLEME RSPQLRKHAC RVMGALNTVV ENLHDPDKVS SVLALVGKAH ALKHKVEPVY FKILSGVILE VVAEEFASDF PPETQRAWAK LRGLIYSHVT AAYKEVGWVQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytoglobin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYGB
Gene Accession No. Q8WWM9
Gene Alias 3110001K20Rik; Cygb; Cytoglobin; HGb; Histoglobin; Staap; STAP; stellate cell activation associated protein; stellate cell activation-associated protein
Gene Symbols CYGB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Cytoglobin. Recombinant protein control fragment (Product #RP-90809).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114757
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.