Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytohesin 1/2 Monoclonal Antibody (6G11)
GREENER_CHOICE

Catalog No. MA1062
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1062 Supplier Invitrogen™ Supplier No. MA1062
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-062 detects recombinant human and mouse cytohesin-1 and 2 (ARNO). MA1-062 has been successfully used in Western blot, IF and immunoprecipitation procedures. By Western blot, this antibody detects an ~48 kDa band representing both cytohesin-1 and cytohesin-2. In immunoprecipitation procedures MA1-062 only precipitates cytohesin-2. The MA1-062 immunogen is purified, His-tagged cytohesin-1.

The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. This gene is highly expressed in natural killer and peripheral T cells, and regulates the adhesiveness of integrins at the plasma membrane of lymphocytes. The encoded protein is 83% homologous to that of CYTH2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 1/2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene CYTH1
Gene Accession No. P63034, Q15438, Q99418, Q9QX11
Gene Alias ARF exchange factor; ARF nucleotide-binding site opener; ARNO; B2-1; B2-1/ARNO; CLM1; CLM2; CTC-273B12.8; CTH-1; CTS18; CTS18.1; Cyth1; Cyth2; CYTIP; cytoadhesin 1; cytohesin 1; cytohesin 2; cytohesin-1; Cytohesin-2; D17S811E; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; mSec7-2; PH, SEC7 and coiled-coil domain-containing protein 1; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled-coil domains 1; pleckstrin homology, Sec7 and coiled-coil domains 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); Protein ARNO; PSCD1; Pscd2; PSCD2L; SEC7; SEC7 homolog A; SEC7 homolog B; SEC7 homolog B2-1; Sec7b; SEC7L; Sec7p-L; Sec7p-like
Gene Symbols CYTH1, CYTH2
Host Species Mouse
Immunogen Purified, His-tagged cytohesin-1.
Purification Method PEG precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19157, 19158, 9266, 9267
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.