Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytohesin-1 Antibody (6G11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222513
Description
Cytohesin-1 Monoclonal specifically detects Cytohesin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Cytohesin-1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:200 - 500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 500 | |
B2-1, cytoadhesin 1, cytohesin 1, cytohesin-1, D17S811ECYTOHESIN-1, FLJ34050, FLJ41900, homolog of secretory protein SEC7, PH, SEC7 and coiled-coil domain-containing protein 1, pleckstrin homology, Sec7 and coiled-coil domains 1, PSCD1, SEC7, SEC7 homolog B2-1 | |
Mouse | |
PEG purified | |
RUO | |
Primary | |
This antibody reacts with Human. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
6G11 | |
Unconjugated | |
Q15438 | |
CYTH1 | |
Purified, His-tagged cytohesin-1. | |
100 μg | |
Membrane Trafficking and Chaperones | |
9267 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction