Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytohesin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590402 Supplier Invitrogen™ Supplier No. PA590402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEDDSYVPS DLTAEERQEL ENIRRRKQEL LADIQRLKDE IAEVANEIEN LGSTEERKNM QRNKQVAMGR; Positive Samples: Raji, Mouse brain, Mouse heart, Mouse spleen, Rat heart; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein, cytosol.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CYTH1
Gene Accession No. P97694, Q15438, Q9QX11
Gene Alias B2-1; CLM1; CTH-1; CYTH1; CYTIP; cytoadhesin 1; cytohesin 1; CYTOHESIN-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; SEC7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a
Gene Symbols CYTH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human CYTH1 (NP_0047531).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116691, 19157, 9267
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.