Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytohesin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584464 Supplier Invitrogen™ Supplier No. PA584464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REELSEAMSE VEGLEANEGS KTLQRNRKMA MGRKKF Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYTH2
Gene Accession No. Q99418
Gene Alias ARF exchange factor; ARF nucleotide binding site opener; ARF nucleotide-binding site opener; ARNO; CLM2; CTC-273B12.8; CTS18; CTS18.1; Cyth2; cytohesin 2; cytohesin-2; mSec7-2; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled-coil domains 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); Protein ARNO; PSCD2; PSCD2L; Sec7; SEC7 homolog B; Sec7b; SEC7L; Sec7p-L; Sec7p-like
Gene Symbols CYTH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYTH2. Recombinant protein control fragment (Product #RP-108448).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.