Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytohesin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102870 Supplier Invitrogen™ Supplier No. PA5102870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CYTH3.

GRP1 (general receptor for phosphoinositides-1) contains a Pleckstrin homology (PH) domain as well as a Sec7 domain. The PH domain has high binding affinity for phosphatidylinositol 3,4,5-trisphosphate Ptdlns(3,4,5)P3), while the Sec7 homology domain is responsible for catalyzing guanine nucleotide exchange of ADP-ribosylation factor (ARF) proteins. GRP1 co-localizes with ARF6 and catalyzes GTP/GDP exchange on ARF6. It is known to interact with Ptdlns(3,4,5)P3 localized to the plasma membrane in vitro and may also be a Ptdlns(3,4,5)P3 receptor. Additionally, GRP1 may regulate protein sorting and membrane trafficking through interaction with the guanosine triphosphate ARF, and may control cell adhesion through interaction with integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cyth3
Gene Accession No. O43739
Gene Alias AI648983; ARF nucleotide-binding site opener 3; ARNO3; CLM3; CYTH3; cytohesin 3; cytohesin-3; General receptor of phosphoinositides 1; GRP1; mSec7-3; PH, SEC7 and coiled-coil domain-containing protein 3; pleckstrin homology, Sec7 and coiled/coil domains 3; pleckstrin homology, Sec7 and coiled-coil domains 3; Protein ARNO3; PSCD3; rSec7-3; Sec7; SEC7 homolog C; Sec7C
Gene Symbols Cyth3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYTH3(Accession O43739), corresponding to amino acid residues G182-P232.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9265
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.