Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytohesin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575368 Supplier Thermo Scientific Supplier No. PA575368
Add to Cart
Add to Cart

Cytohesin 3 Polyclonal Antibody

Cytohesin 3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Cyth3
Gene Accession No. O08967, O43739, P97696
Gene Alias AI648983; ARF nucleotide-binding site opener 3; ARNO3; CLM3; CYTH3; cytohesin 3; cytohesin-3; General receptor of phosphoinositides 1; GRP1; mSec7-3; PH, SEC7 and coiled-coil domain-containing protein 3; pleckstrin homology, Sec7 and coiled/coil domains 3; pleckstrin homology, Sec7 and coiled-coil domains 3; Protein ARNO3; PSCD3; rSec7-3; Sec7; SEC7 homolog C; Sec7C
Gene Symbols Cyth3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 371-400 of Human GRP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116693, 19159, 9265
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.