Learn More
Invitrogen™ Cytohesin 3 Polyclonal Antibody

Description
Immunogen sequence: DEDGGGEGGG VPEDLSLEER EELLDIRRRK KELIDDIERL KYEIAEVMTE IDNLTSVEES KTTQRNKQIA M Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 86%.
Specifications
Specifications
| Antigen | Cytohesin 3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Cyth3 |
| Gene Accession No. | O43739 |
| Gene Alias | AI648983; ARF nucleotide-binding site opener 3; ARNO3; CLM3; CYTH3; cytohesin 3; cytohesin-3; General receptor of phosphoinositides 1; GRP1; mSec7-3; PH, SEC7 and coiled-coil domain-containing protein 3; pleckstrin homology, Sec7 and coiled/coil domains 3; pleckstrin homology, Sec7 and coiled-coil domains 3; Protein ARNO3; PSCD3; rSec7-3; Sec7; SEC7 homolog C; Sec7C |
| Gene Symbols | Cyth3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.