Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cytohesin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564467
Description
Immunogen sequence: MDLCHPEPAE LSSGETEELQ RIKWHRKQLL EDIQKL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with both ARF1 and ARF5 but is inactive with ARF6. The structures of this gene and CYTH1 are very similar.
Specifications
Cytohesin 4 | |
Polyclonal | |
Unconjugated | |
CYTH4 | |
CYT4; CYTH4; cytohesin 4; cytohesin-4; DJ63G5.1; PH, SEC7 and coiled-coil domain-containing protein 4; pleckstrin homology, Sec7 and coiled/coil domains 4; pleckstrin homology, Sec7 and coiled-coil domains 4; PSCD4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27128 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIA0 | |
CYTH4 | |
Recombinant protein corresponding to Human Cytohesin 4. Recombinant protein control fragment (Product #RP-104282). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction