Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 1 Antibody (3C10F7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11089450
Description
Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Porcine, Bovine, Canine, Equine, Feline, Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.Specifications
Cytokeratin 1 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
KRT1 | |
Ni-NTA purified truncated recombinant CK1 expressed in E. Coli strain BL21 (DE3) | |
0.1 mL | |
Protein Kinase | |
3848 | |
Human, Pig, Bovine, Canine, Equine, Feline, Goat | |
Ascites |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
3C10F7 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry Free-Floating | |
CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
CK1 (PMC01) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction