Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298734H
Description
Cytokeratin 1 Polyclonal specifically detects Cytokeratin 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cytokeratin 1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Cytokeratin 1. | |
0.1 mL | |
Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction | |
3848 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction