Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 1 Mouse anti-Human, Rat, Clone: LHK1, Novus Biologicals™
SDP

Catalog No. p-7232974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123024 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123024 Supplier Novus Biologicals Supplier No. NBP30777520UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Rat samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen Cytokeratin 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LHK1
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.1-0.2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1
Host Species Mouse
Immunogen A 13-amino acid peptide (VRFVSTSYSGVTR) from the C-terminus of human Cytokeratin 1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3848
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.