Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytokeratin 1 Antibody (LHK1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280688
Description
Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Cytokeratin 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5-2 μg/mL, Immunohistochemistry : 2-10 μg/mL, Immunocytochemistry/Immunofluorescence : 5 μg/mL, Immunohistochemistry-Paraffin : 2-5 μg/mL, Immunohistochemistry-Frozen : 5-10 μg/mL | |
CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
LHK1 | |
Unconjugated | |
PBS | |
KRT1 | |
C-terminal peptide sequence of human Cytokeratin 1 [UniProt# P04264] | |
0.1 mL | |
Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction | |
3848 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction